Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam115e
Entrez Gene ID
494321
Gene Name
family with sequence similarity 115, member E
Gene Aliases
Eapa2
Chromosome Location
Chr.4: 72010583 - 72023952 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009534.1 NP_001009534.1 2 - 3 674 94
GenBank mRNA BC061995.1 - 2 - 3 674 94

View Full Product Details