The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004086.1 | NP_001004086.1 | 6 - 7 | 701 | 137 |
GenBank mRNA | BC079466.1 | - | 6 - 7 | 701 | 137 |
FQ218352.1 | - | 3 - 4 | 440 | 137 | |
FQ218880.1 | - | 3 - 4 | 440 | 137 | |
FQ218895.1 | - | 3 - 4 | 441 | 137 | |
FQ219524.1 | - | 3 - 4 | 440 | 137 |