The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002808.2 | NP_001002808.2 | 5 - 6 | 950 | 66 |
XM_006236215.2 | XP_006236277.1 | 6 - 7 | 1489 | 66 | |
XM_006236216.2 | XP_006236278.1 | 6 - 7 | 1213 | 66 | |
XM_006236217.2 | XP_006236279.1 | 6 - 7 | 1173 | 66 | |
XM_006236218.2 | XP_006236280.1 | 6 - 7 | 744 | 66 | |
XM_006236219.2 | XP_006236281.1 | 5 - 6 | 1638 | 66 | |
XM_008762801.1 | XP_008761023.1 | 6 - 7 | 1529 | 66 | |
XM_008762802.1 | XP_008761024.1 | 6 - 7 | 1505 | 66 | |
XM_008762803.1 | XP_008761025.1 | 5 - 6 | 1638 | 66 | |
GenBank mRNA | BC085840.1 | - | 5 - 6 | 950 | 66 |