Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gal
Entrez Gene ID
29141
Gene Name
galanin/GMAP prepropeptide
Gene Aliases
Galn
Chromosome Location
Chr.1: 218653059 - 218657922 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033237.1 NP_150240.1 2 - 3 223 61
GenBank mRNA J03624.1 - 2 - 3 223 61
M18102.1 - 2 - 3 203 61

View Full Product Details