Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sept3
Entrez Gene ID
56003
Gene Name
septin 3
Gene Aliases
Sep3
Chromosome Location
Chr.7: 123506535 - 123526542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019375.1 NP_062248.1 5 - 6 575 84
GenBank mRNA AF111179.1 - 5 - 6 575 84
AF111180.1 - 3 - 4 982 84
AF111181.1 - 5 - 6 574 84

View Full Product Details