Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trps1
Entrez Gene ID
299897
Gene Name
trichorhinophalangeal syndrome I
Gene Aliases
-
Chromosome Location
Chr.7: 90090828 - 90318748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134837.1 NP_001128309.1 3 - 4 1120 75
XM_008765496.1 XP_008763718.1 2 - 3 1309 75
XM_008765497.1 XP_008763719.1 2 - 3 2130 75
XM_008765498.1 XP_008763720.1 2 - 3 974 75
XM_008765499.1 XP_008763721.1 2 - 3 1309 75

View Full Product Details