Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnq3
Entrez Gene ID
29682
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 3
Gene Aliases
-
Chromosome Location
Chr.7: 106714479 - 107009639 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031597.4 NP_113785.3 5 - 6 1360 89
XM_008765495.1 XP_008763717.1 5 - 6 688 89
GenBank mRNA AF091247.1 - 6 - 7 906 89

View Full Product Details