Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc39a4
Entrez Gene ID
300051
Gene Name
solute carrier family 39 (zinc transporter), member 4
Gene Aliases
-
Chromosome Location
Chr.7: 117675718 - 117682580 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077669.1 NP_001071137.1 10 - 11 1751 111
XM_008765563.1 XP_008763785.1 11 - 12 3928 111
XM_008765564.1 XP_008763786.1 11 - 12 3806 111
GenBank mRNA BC127514.1 - 10 - 11 1751 111

View Full Product Details