Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gsdmc
Entrez Gene ID
299908
Gene Name
gasdermin C
Gene Aliases
Gsdmc1, Mlze
Chromosome Location
Chr.7: 104474418 - 104492364 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134495.1 NP_001127967.1 9 - 10 1094 125
XM_006241631.2 XP_006241693.1 13 - 14 1758 125

View Full Product Details