The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013948.1 | NP_001013970.1 | 4 - 5 | 675 | 88 |
XM_006241530.2 | XP_006241592.1 | 4 - 5 | 946 | 88 | |
XM_006241531.2 | XP_006241593.1 | 3 - 4 | 578 | 88 | |
XM_006241532.2 | XP_006241594.1 | 3 - 4 | 2319 | 88 | |
XM_006241533.2 | XP_006241595.1 | 3 - 4 | 2245 | 88 | |
GenBank mRNA | AB095364.1 | - | 4 - 5 | 675 | 88 |
BC105616.1 | - | 4 - 5 | 671 | 88 |