Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eri2
Entrez Gene ID
691484
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
Exod1
Chromosome Location
Chr.1: 189540145 - 189549891 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191114.1 NP_001178043.1 3 - 4 181 71

View Full Product Details