Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olr59
Entrez Gene ID
170816
Gene Name
olfactory receptor 59
Gene Aliases
Olfr78, Or51e2, RA1c
Chromosome Location
Chr.1: 167906030 - 167940688 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759669.1 XP_008757891.1 1 - 2 1899 129

View Full Product Details