Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmtc3
Entrez Gene ID
314785
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
RGD1306351
Chromosome Location
Chr.7: 40171957 - 40217056 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135858.1 NP_001129330.1 10 - 11 1606 150
XM_008765316.1 XP_008763538.1 9 - 10 1516 150

View Full Product Details