The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135858.1 | NP_001129330.1 | 14 - 15 | 2224 | 92 |
XM_008765316.1 | XP_008763538.1 | 13 - 14 | 2134 | 92 |