The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021745.1 | NP_068513.1 | 8 - 9 | 1228 | 90 |
XM_008765260.1 | XP_008763482.1 | 7 - 8 | 1598 | 90 | |
XM_008765261.1 | XP_008763483.1 | 7 - 8 | 1585 | 90 | |
XM_008765262.1 | XP_008763484.1 | 8 - 9 | 1279 | 90 | |
GenBank mRNA | BC083815.1 | - | 7 - 8 | 1106 | 90 |
U18374.1 | - | 8 - 9 | 1228 | 90 |