The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108100.1 | NP_001101570.1 | 1 - 2 | 62 | 63 |
XM_006241390.2 | XP_006241452.1 | 2 - 3 | 257 | 63 | |
XM_006241391.2 | XP_006241453.1 | 2 - 3 | 277 | 63 | |
XM_008765401.1 | XP_008763623.1 | 2 - 3 | 667 | 63 |