Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc90b
Entrez Gene ID
308820
Gene Name
coiled-coil domain containing 90B
Gene Aliases
RGD1308637
Chromosome Location
Chr.1: 157403595 - 157417153 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024885.1 NP_001020056.1 6 - 7 846 74
GenBank mRNA BC097480.1 - 6 - 7 846 74
FQ215669.1 - 6 - 7 548 74
FQ218235.1 - 6 - 7 561 74

View Full Product Details