Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx6
Entrez Gene ID
365371
Gene Name
T-box 6
Gene Aliases
-
Chromosome Location
Chr.1: 198214797 - 198218706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108920.1 NP_001102390.1 3 - 4 646 117

View Full Product Details