Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp35
Entrez Gene ID
308834
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
RGD1565984
Chromosome Location
Chr.1: 162283866 - 162307604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759807.1 XP_008758029.1 5 - 6 1558 97
XM_238899.7 XP_238899.4 4 - 5 1361 97

View Full Product Details