Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Calca
Entrez Gene ID
24241
Gene Name
calcitonin-related polypeptide alpha
Gene Aliases
CAL6, CGRP, Cal1, Calc, RATCAL6, calcitonin
Chromosome Location
Chr.1: 184184018 - 184188938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033955.1 NP_001029127.1 3 - 4 358 69
NM_001033956.1 NP_001029128.1 3 - 4 334 69
GenBank mRNA BG663259.1 - 3 - 4 300 69
CB711853.1 - 3 - 4 326 69
CF978038.1 - 3 - 4 339 69
M11597.1 - 2 - 3 232 69
V01231.1 - 2 - 3 231 69

View Full Product Details