Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdr2
Entrez Gene ID
308958
Gene Name
cerebellar degeneration-related protein 2
Gene Aliases
RGD1310578
Chromosome Location
Chr.1: 190889698 - 190914606 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025682.1 NP_001020853.1 3 - 4 410 102
GenBank mRNA BC098028.1 - 3 - 4 410 102

View Full Product Details