Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bysl
Entrez Gene ID
359727
Gene Name
bystin-like
Gene Aliases
-
Chromosome Location
Chr.9: 15393461 - 15403212 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182674.3 NP_872615.3 1 - 2 333 106
GenBank mRNA AY257675.4 - 1 - 2 316 106
BC079030.1 - 1 - 2 322 106
CV104390.1 - 1 - 2 333 106

View Full Product Details