Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bai3
Entrez Gene ID
301309
Gene Name
brain-specific angiogenesis inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.9: 31280623 - 32022098 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106898.1 NP_001100368.1 19 - 20 3029 156
XM_008766928.1 XP_008765150.1 3 - 4 720 156
XM_008766929.1 XP_008765151.1 4 - 5 613 156
XM_008766930.1 XP_008765152.1 4 - 5 2034 156
XM_008766931.1 XP_008765153.1 4 - 5 850 156
XM_008766932.1 XP_008765154.1 3 - 4 720 156

View Full Product Details