Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cops2
Entrez Gene ID
261736
Gene Name
COP9 signalosome subunit 2
Gene Aliases
CSN2, Trip15
Chromosome Location
Chr.3: 118109394 - 118137125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153297.1 NP_695209.1 8 - 9 1046 168
XM_006234901.2 XP_006234963.1 8 - 9 996 168
XM_008762146.1 XP_008760368.1 8 - 9 852 168
GenBank mRNA AB081072.1 - 8 - 9 1046 168
BC061864.1 - 8 - 9 823 168
FQ210124.1 - 8 - 9 971 168

View Full Product Details