Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Metap1d
Entrez Gene ID
311748
Gene Name
methionyl aminopeptidase type 1D (mitochondrial)
Gene Aliases
Map1d, Metapl1
Chromosome Location
Chr.3: 58084558 - 58163259 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107812.2 NP_001101282.2 1 - 2 103 92
XM_008761908.1 XP_008760130.1 1 - 2 98 92
GenBank mRNA DY575699.1 - 1 - 2 130 92

View Full Product Details