Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmx4
Entrez Gene ID
296182
Gene Name
thioredoxin-related transmembrane protein 4
Gene Aliases
RGD1305146, Txndc13
Chromosome Location
Chr.3: 127528427 - 127569093 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100529.1 NP_001093999.1 4 - 5 596 125

View Full Product Details