The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100771.1 | NP_001094241.1 | 13 - 14 | 2357 | 108 |
XM_006235105.2 | XP_006235167.1 | 13 - 14 | 2490 | 108 | |
XM_006235106.2 | XP_006235168.1 | 13 - 14 | 2356 | 108 | |
GenBank mRNA | AB038233.1 | - | 13 - 14 | 2355 | 108 |