The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048184.1 | NP_001041649.1 | 5 - 6 | 938 | 99 |
GenBank mRNA | BC105622.1 | - | 5 - 6 | 938 | 99 |