Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35e2b
Entrez Gene ID
313765
Gene Name
solute carrier family 35, member E2B
Gene Aliases
Slc35e2
Chromosome Location
Chr.5: 173024335 - 173050228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107998.1 NP_001101468.1 8 - 9 1882 69
XM_006239572.2 XP_006239634.1 8 - 9 1444 69

View Full Product Details