Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc27
Entrez Gene ID
679584
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
Chromosome Location
Chr.5: 171312790 - 171327581 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191063.1 NP_001177992.1 11 - 12 1747 96
XM_008764384.1 XP_008762606.1 12 - 13 1911 96

View Full Product Details