Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dvl1
Entrez Gene ID
83721
Gene Name
dishevelled segment polarity protein 1
Gene Aliases
dvl-1
Chromosome Location
Chr.5: 173295955 - 173308014 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031820.1 NP_114008.1 8 - 9 908 61
XM_006239598.2 XP_006239660.1 8 - 9 1222 61
XM_008764388.1 XP_008762610.1 7 - 8 1310 61
XM_008764389.1 XP_008762611.1 7 - 8 1310 61
XM_008764390.1 XP_008762612.1 7 - 8 1310 61
XM_008764391.1 XP_008762613.1 8 - 9 1011 61
GenBank mRNA AF143545.2 - 8 - 9 908 61
AF143546.2 - 8 - 9 908 61
BC160920.1 - 1 - 2 57 61

View Full Product Details