Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina11
Entrez Gene ID
362774
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11
Gene Aliases
-
Chromosome Location
Chr.6: 127647180 - 127656603 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166352.1 NP_001159824.1 3 - 4 1022 68
GenBank mRNA BC111708.1 - 2 - 3 934 68
FQ211426.1 - 3 - 4 994 68

View Full Product Details