The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008776.2 | NP_001008776.1 | 4 - 5 | 1179 | 74 |
NM_001166352.1 | NP_001159824.1 | 4 - 5 | 1165 | 74 | |
XM_006240481.2 | XP_006240543.1 | 5 - 6 | 1284 | 74 | |
GenBank mRNA | AY325135.1 | - | 4 - 5 | 1219 | 74 |
BC111708.1 | - | 3 - 4 | 1077 | 74 | |
FQ211426.1 | - | 4 - 5 | 1137 | 74 |