Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psmc1
Entrez Gene ID
117263
Gene Name
proteasome (prosome, macropain) 26S subunit, ATPase, 1
Gene Aliases
s4
Chromosome Location
Chr.6: 124123283 - 124135644 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057123.1 NP_476464.1 1 - 2 67 80
GenBank mRNA BC063157.1 - 2 - 3 79 80
D50696.1 - 1 - 2 67 80
FQ209916.1 - 2 - 3 72 80

View Full Product Details