Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gar1
Entrez Gene ID
499709
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
Nola1
Chromosome Location
Chr.2: 235250453 - 235258460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024306.1 NP_001019477.1 6 - 7 932 107
GenBank mRNA BC079131.1 - 6 - 7 932 107

View Full Product Details