Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfap2
Entrez Gene ID
313662
Gene Name
microfibrillar-associated protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 159534202 - 159539750 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107989.1 NP_001101459.1 2 - 3 117 69

View Full Product Details