Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkcz
Entrez Gene ID
25522
Gene Name
protein kinase C, zeta
Gene Aliases
14-3-3-zetaisoform, Pkcz, r14-3-3
Chromosome Location
Chr.5: 172658071 - 172769492 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022507.1 NP_071952.1 14 - 15 1548 91
XM_006239535.2 XP_006239597.1 11 - 12 4043 91
XM_006239536.2 XP_006239598.1 11 - 12 3249 91
XM_008764337.1 XP_008762559.1 14 - 15 1449 91
GenBank mRNA J04532.1 - 14 - 15 1548 91
M18332.1 - 11 - 12 1101 91
U27191.1 - 11 - 12 1313 91

View Full Product Details