Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrto4
Entrez Gene ID
298586
Gene Name
mRNA turnover 4 homolog (S. cerevisiae)
Gene Aliases
RGD1311709
Chromosome Location
Chr.5: 157814142 - 157820889 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106697.1 NP_001100167.1 7 - 8 997 87
XM_006239178.2 XP_006239240.1 7 - 8 865 87
XM_008764230.1 XP_008762452.1 6 - 7 599 87
GenBank mRNA BC168898.1 - 7 - 8 564 87
FQ227354.1 - 7 - 8 610 87
FQ227472.1 - 7 - 8 849 87
FQ234412.1 - 7 - 8 849 87

View Full Product Details