The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029923.1 | NP_001025094.1 | 9 - 10 | 1068 | 77 |
GenBank mRNA | AB299031.1 | - | 9 - 10 | 971 | 77 |
BC098048.1 | - | 9 - 10 | 1068 | 77 |