Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ero1lb
Entrez Gene ID
364755
Gene Name
ERO1-like beta (S. cerevisiae)
Gene Aliases
RGD1563548
Chromosome Location
Chr.17: 90709394 - 90781848 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191893.1 NP_001178822.1 3 - 4 311 107
XM_006254375.2 XP_006254437.1 3 - 4 501 107
XM_008771876.1 XP_008770098.1 3 - 4 277 107

View Full Product Details