Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet2
Entrez Gene ID
310859
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
RGD1311625
Chromosome Location
Chr.2: 238719389 - 238802979 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233347.2 XP_006233409.1 2 - 3 313 100
XM_008761534.1 XP_008759756.1 2 - 3 313 100
XM_227694.7 XP_227694.5 2 - 3 313 100

View Full Product Details