Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rtcd1
Entrez Gene ID
295395
Gene Name
RNA terminal phosphate cyclase domain 1
Gene Aliases
-
Chromosome Location
Chr.2: 219537213 - 219558489 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004227.1 NP_001004227.1 4 - 5 424 104
XM_008761456.1 XP_008759678.1 3 - 4 380 104
GenBank mRNA BC079374.1 - 4 - 5 424 104

View Full Product Details