Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndst4
Entrez Gene ID
362035
Gene Name
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4
Gene Aliases
-
Chromosome Location
Chr.2: 229195261 - 229496732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191849.1 NP_001178778.1 7 - 8 1814 106
XM_006233274.2 XP_006233336.1 8 - 9 2135 106

View Full Product Details