Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints12
Entrez Gene ID
295448
Gene Name
integrator complex subunit 12
Gene Aliases
Phf22
Chromosome Location
Chr.2: 238253119 - 238270722 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233321.2 XP_006233383.1 1 - 2 116 92
GenBank mRNA FQ212179.1 - 1 - 2 91 92

View Full Product Details