Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd13c
Entrez Gene ID
685374
Gene Name
ankyrin repeat domain 13C
Gene Aliases
-
Chromosome Location
Chr.2: 264785380 - 264833283 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191570.1 NP_001178499.1 11 - 12 1521 95
XM_006233547.2 XP_006233609.1 10 - 11 2446 95
XM_008761571.1 XP_008759793.1 9 - 10 2342 95

View Full Product Details