The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012059.1 | NP_001012059.1 | 12 - 13 | 1680 | 94 |
GenBank mRNA | BC079200.1 | - | 12 - 13 | 1680 | 94 |