Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aurkc
Entrez Gene ID
292554
Gene Name
aurora kinase C
Gene Aliases
-
Chromosome Location
Chr.1: 69958452 - 69964105 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106221.1 NP_001099691.1 1 - 2 151 107
XM_008758900.1 XP_008757122.1 4 - 5 515 107
XM_008758901.1 XP_008757123.1 5 - 6 618 107

View Full Product Details