The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022513.2 | NP_071958.1 | 4 - 5 | 594 | 92 |
XM_008770056.1 | XP_008768278.1 | 4 - 5 | 468 | 92 | |
GenBank mRNA | D89375.1 | - | 4 - 5 | 545 | 92 |
FQ209476.1 | - | 4 - 5 | 562 | 92 | |
FQ209486.1 | - | 4 - 5 | 628 | 92 | |
FQ209579.1 | - | 4 - 5 | 649 | 92 | |
FQ210801.1 | - | 4 - 5 | 636 | 92 | |
FQ212608.1 | - | 4 - 5 | 605 | 92 | |
U38419.1 | - | 4 - 5 | 597 | 92 |