The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173106.1 | NP_775129.1 | 8 - 9 | 555 | 104 |
GenBank mRNA | AY154894.1 | - | 8 - 9 | 555 | 104 |