Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zkscan4
Entrez Gene ID
291164
Gene Name
zinc finger with KRAB and SCAN domains 4
Gene Aliases
Zfp307, Znf307
Chromosome Location
Chr.17: 45147545 - 45154355 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254110.2 XP_006254172.2 2 - 3 323 68

View Full Product Details